L-Glutamine-13C5,15N2
Synonym(s):
(S)-2,5-Diamino-5-oxopentanoic acid-13C5-15N2, L-Glutamic acid 5-amide-13C5-15N2, L-Glutamine-ul-13C,15N, 13C and 15N Labeled glutamine
Linear Formula:
H215N13CO(13CH2)213CH(15NH2)13CO2H
Molecular Weight:
153.09
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing