Levofloxacin Related Compound E
Synonym(s):
(3S)-9-Fluoro-2,3-dihydro-3-methyl-10-[2-(methylamino)ethylamino]-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid monohydrochloride
Empirical Formula (Hill Notation):
C16H18FN3O4 · HCl
CAS Number:
Molecular Weight:
371.79
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2