![Levetiracetam-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/160/593/26a4c35c-41bb-47d3-b10a-936bb4a48910/640/26a4c35c-41bb-47d3-b10a-936bb4a48910.png)
Levetiracetam-D6 solution
![Levetiracetam-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/160/593/26a4c35c-41bb-47d3-b10a-936bb4a48910/640/26a4c35c-41bb-47d3-b10a-936bb4a48910.png)
Synonym(s):
Levetiracetam-D6 solution
Empirical Formula (Hill Notation):
C8D6H8N2O2
Molecular Weight:
176.25
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique