Skip to Content
Merck
Leuprolide acetate salt ≥98% (HPLC)

Leuprolide acetate salt

Leuprolide acetate salt ≥98% (HPLC)
Synonym(s):
des-Gly10-[D-Leu6]-LH-RH ethylamide, Leuprorelin
Empirical Formula (Hill Notation):
C59H84N16O12 · xC2H4O2
CAS Number:
Molecular Weight:
1209.40 (free base basis)

Products

Brand
Formula Weight
Gene Alias
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing