Skip to Content
Merck
[Leu15]-Gastrin I human ≥95% (HPLC)

[Leu15]-Gastrin I human

[Leu15]-Gastrin I human ≥95% (HPLC)
Synonym(s):
[Leu15]-HG-17, LHG-17, [15-Leucine]-Gastrin I, human heptadecapeptide, [18-Pyroglutamic acid, 32-leucine]-Big gastrin I-(18-34)-peptide amide, human, [Leu15]-Gastrin I, human, [Leucine15]-Gastrin I heptadecapeptide, [Leucine15]-Little gastrin I
Empirical Formula (Hill Notation):
C98H126N20O31
CAS Number:
Molecular Weight:
2080.16
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing