Letrozole impurity A
Synonym(s):
4,4′-(4H-1,2,4-Triazol-4-ylmethylene)bisbenzonitrile, 4,4′-(1H-1,3,4-Triazol-1-ylmethylene)dibenzonitrile
Empirical Formula (Hill Notation):
C17H11N5
CAS Number:
Molecular Weight:
285.30
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing