L-(+)-2-Amino-3-phosphonopropionic acid
Synonym(s):
(R)-(+)-2-Amino-3-phosphonopropionic acid, 3-Phosphono-L-alanine, L-AP3
Empirical Formula (Hill Notation):
C3H8NO5P
CAS Number:
Molecular Weight:
169.07
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing