![L-Cysteine-13C3,15N ≥99 atom %, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/143/163/8d8aaab6-a7aa-4fe4-9e4b-5f5b1a891a8e/640/8d8aaab6-a7aa-4fe4-9e4b-5f5b1a891a8e.png)
L-Cysteine-13C3,15N
![L-Cysteine-13C3,15N ≥99 atom %, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/143/163/8d8aaab6-a7aa-4fe4-9e4b-5f5b1a891a8e/640/8d8aaab6-a7aa-4fe4-9e4b-5f5b1a891a8e.png)
Synonym(s):
(R)-2-Amino-3-mercaptopropionic acid-13C3,15N, 13C and 15N Labeled L-cysteine
Linear Formula:
HS13CH213CH(15NH2)13COOH
CAS Number:
Molecular Weight:
125.13
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing