![L-Arginine, Hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/391/654/983d7f2d-d2ea-4d17-b3e3-bb38f229592d/640/983d7f2d-d2ea-4d17-b3e3-bb38f229592d.png)
L-Arginine, Hydrochloride
![L-Arginine, Hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/391/654/983d7f2d-d2ea-4d17-b3e3-bb38f229592d/640/983d7f2d-d2ea-4d17-b3e3-bb38f229592d.png)
Synonym(s):
L-Arginine monohydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride
Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing