![Lamivudine for system suitability 2 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/419/898/e611d53e-454d-4c8a-8614-9a54edafe198/640/e611d53e-454d-4c8a-8614-9a54edafe198.png)
Lamivudine for system suitability 2
![Lamivudine for system suitability 2 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/419/898/e611d53e-454d-4c8a-8614-9a54edafe198/640/e611d53e-454d-4c8a-8614-9a54edafe198.png)
Synonym(s):
Lamivudine, 2′,3′-Dideoxy-3′-thiacytidine, 3TC, Epivir, Epivir-HBV, Heptovir, Zeffix
Empirical Formula (Hill Notation):
C8H11N3O3S
CAS Number:
Molecular Weight:
229.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing