Lactose, Anhydrous
Synonym(s):
Lactose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Analyte Suitability
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2