![Kaempferol 3-sambubioside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/121/539/af7ac5e6-d751-4f6e-817e-1577cd9b1aea/640/af7ac5e6-d751-4f6e-817e-1577cd9b1aea.png)
Kaempferol 3-sambubioside
![Kaempferol 3-sambubioside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/121/539/af7ac5e6-d751-4f6e-817e-1577cd9b1aea/640/af7ac5e6-d751-4f6e-817e-1577cd9b1aea.png)
Synonym(s):
Leucoside, Kaempferol 3-sambubioside
Empirical Formula (Hill Notation):
C26H28O15
CAS Number:
Molecular Weight:
580.49
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing