![Isradipine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/252/210/7b9b0337-4aaa-4855-b482-f209f712f10d/640/7b9b0337-4aaa-4855-b482-f209f712f10d.png)
Isradipine
![Isradipine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/252/210/7b9b0337-4aaa-4855-b482-f209f712f10d/640/7b9b0337-4aaa-4855-b482-f209f712f10d.png)
Synonym(s):
4-(4-Benzofurazanyl)-1,-4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid methyl 1-methylethyl ester, Isradipine
Empirical Formula (Hill Notation):
C19H21N3O5
CAS Number:
Molecular Weight:
371.39
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4