Skip to Content
Merck
Isosilybin ≥98% (HPLC)

Isosilybin

Isosilybin ≥98% (HPLC)
Synonym(s):
Isosilybin, mixture of isomers, Isosilybinin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing