![Isoscopoletin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/412/435/e90b106e-3bbf-4e1f-a984-9c758d982b13/640/e90b106e-3bbf-4e1f-a984-9c758d982b13.png)
Isoscopoletin
![Isoscopoletin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/412/435/e90b106e-3bbf-4e1f-a984-9c758d982b13/640/e90b106e-3bbf-4e1f-a984-9c758d982b13.png)
Synonym(s):
6-Hydroxy-7-methoxycoumarin, 7-Methoxyesculetin, Esculetin 7-methyl ether, Isoscopoletin
Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing