Isobutyl chloroformate
Synonym(s):
Chloroformic acid isobutyl ester, IBCF, Isobutyl chloroformate
Linear Formula:
ClCOOCH2CH(CH3)2
CAS Number:
Molecular Weight:
136.58
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing