Skip to Content
Merck
Isatin 97%

Isatin

Isatin 97%
Synonym(s):
2,3-Indolinedione, Isatine, NSC 9262, Isatin
Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing