Skip to Content
Merck
Insulin from bovine pancreas powder, BioReagent, suitable for cell culture

Insulin from bovine pancreas

Insulin from bovine pancreas powder, BioReagent, suitable for cell culture
Empirical Formula (Hill Notation):
C254H377N65O75S6
CAS Number:
Molecular Weight:
5733.49
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique