![Indole-3-acetaldehyde–sodium bisulfite addition compound](/deepweb/assets/sigmaaldrich/product/structures/191/855/7f04497a-5b21-489c-b82c-b006dc33a3f0/640/7f04497a-5b21-489c-b82c-b006dc33a3f0.png)
Indole-3-acetaldehyde–sodium bisulfite addition compound
![Indole-3-acetaldehyde–sodium bisulfite addition compound](/deepweb/assets/sigmaaldrich/product/structures/191/855/7f04497a-5b21-489c-b82c-b006dc33a3f0/640/7f04497a-5b21-489c-b82c-b006dc33a3f0.png)
Linear Formula:
C10H9NO · NaHSO3
CAS Number:
Molecular Weight:
263.25
Products
Product Category
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing