Skip to Content
Merck
[Hydroxy(tosyloxy)iodo]benzene 96%

[Hydroxy(tosyloxy)iodo]benzene

[Hydroxy(tosyloxy)iodo]benzene 96%
Synonym(s):
HTIB, Hydroxy(4-methylbenzenesulfonato-O)phenyliodine, Hydroxy(phenyl)iodo tosylate, Iodosobenzene-I-mono-p-toluenesulfonate, Koser’s reagent, NSC 294176, Phenyliodosyl hydroxide tosylate, [Hydroxy(4-toluenesulfonato)iodo]benzene
Linear Formula:
CH3C6H4SO3I(OH)C6H5
CAS Number:
Molecular Weight:
392.21

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing