![Hydrocortisone sodium phosphate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/004/539e7c01-3254-43d8-b39b-0e43e328f5f9/640/539e7c01-3254-43d8-b39b-0e43e328f5f9.png)
Hydrocortisone sodium phosphate
![Hydrocortisone sodium phosphate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/004/539e7c01-3254-43d8-b39b-0e43e328f5f9/640/539e7c01-3254-43d8-b39b-0e43e328f5f9.png)
Synonym(s):
Hydrocortisone 21-phosphate sodium salt, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-phosphate, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-phosphate, Cortisol 21-phosphate sodium salt
Empirical Formula (Hill Notation):
C21H29Na2O8P
CAS Number:
Molecular Weight:
486.40
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing