Skip to Content
Merck
H-Lys(Z)-OMe hydrochloride ≥97.5% (HPLC)

H-Lys(Z)-OMe hydrochloride

H-Lys(Z)-OMe hydrochloride ≥97.5% (HPLC)
Synonym(s):
Nε-Z-L-lysine methyl ester hydrochloride
Linear Formula:
C6H5CH2OOCNH(CH2)4CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
330.81
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing