Skip to Content
Merck
Hederacoside C primary reference standard

Hederacoside C

Hederacoside C primary reference standard
Empirical Formula (Hill Notation):
C59H96O26
CAS Number:
Molecular Weight:
1221.38
EC Number:

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3