Glipizide impurity D
Synonym(s):
6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
Empirical Formula (Hill Notation):
C14H16N4O3S
CAS Number:
Molecular Weight:
320.37
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing