Skip to Content
Merck
Glabridin phyproof® Reference Substance

Glabridin

Glabridin phyproof® Reference Substance
Synonym(s):
(R)-4-(3,4-Dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b′]dipyran-3-yl)-1,3-benzenediol
Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing