Skip to Content
Merck
Ganoderic acid D phyproof® Reference Substance

Ganoderic acid D

Ganoderic acid D phyproof® Reference Substance
Synonym(s):
(7β)-7-Hydroxy-3,11,15,23-tetraoxolanost-8-en-26-oic acid, GAD
Empirical Formula (Hill Notation):
C30H42O7
CAS Number:
Molecular Weight:
514.65

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing