Skip to Content
Merck
Gabexate mesylate ≥97% (HPLC)

Gabexate mesylate

Gabexate mesylate ≥97% (HPLC)
Synonym(s):
Gabexate mesylate, 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxyl]benzoic acid ethyl ester mesylate salt, FOY, Megacart
Empirical Formula (Hill Notation):
C16H23N3O4 · CH4O3S
CAS Number:
Molecular Weight:
417.48

Products

Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing