
Fmoc-Asp-OtBu

Synonym(s):
Fmoc-Asp-OtBu
Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing