Skip to Content
Merck
Fmoc-Asn(Trt)-OH ≥97.0%

Fmoc-Asn(Trt)-OH

Fmoc-Asn(Trt)-OH ≥97.0%
Synonym(s):
Nα-(9-Fluorenylmethoxycarbonyl)-Nγ-trityl-L-asparagine, Nα-Fmoc-Nγ-trityl-L-asparagine, FMOC-Asn(Trt)-OH
Empirical Formula (Hill Notation):
C38H32N2O5
CAS Number:
Molecular Weight:
596.67

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2