
Fmoc-Ala-OH-3-13C

Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-alanine-3-13C, L-Alanine-3-13C, N-Fmoc
Linear Formula:
13CH3CH(NH-Fmoc)CO2H
CAS Number:
Molecular Weight:
312.32
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1