Skip to Content
Merck
Flutamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Flutamide

Flutamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Empirical Formula (Hill Notation):
C11H11F3N2O3
CAS Number:
Molecular Weight:
276.21
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Product Line
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade