![Fluopyram PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/185/249/0e97b800-5af5-4793-a2d6-610efc388f19/640/0e97b800-5af5-4793-a2d6-610efc388f19.png)
Fluopyram
![Fluopyram PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/185/249/0e97b800-5af5-4793-a2d6-610efc388f19/640/0e97b800-5af5-4793-a2d6-610efc388f19.png)
Synonym(s):
N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide, N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-α,α,α-trifluoro-o-toluamide
Empirical Formula (Hill Notation):
C16H11ClF6N2O
CAS Number:
Molecular Weight:
396.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2