Skip to Content
Merck
Flumethasone pivalate European Pharmacopoeia (EP) Reference Standard

Flumethasone pivalate

Flumethasone pivalate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
6α,9-Difluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-(2,2-dimethylpropionate), Flumetasone 21-pivalate, NSC 107680
Empirical Formula (Hill Notation):
C27H36F2O6
CAS Number:
Molecular Weight:
494.57

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade