Fingolimod Hydrochloride
Synonym(s):
FTY720, 2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride
Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
Products
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3