Fenoldopam mesylate
Synonym(s):
Fenoldopam methanesulfonate
Empirical Formula (Hill Notation):
C16H16ClNO3 · CH4O3S
CAS Number:
Molecular Weight:
401.86
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing