![Fast Blue RR Salt Preweighed Capsules](/deepweb/assets/sigmaaldrich/product/structures/114/647/816c95a3-8172-4ea0-b156-e71a1e87cc45/640/816c95a3-8172-4ea0-b156-e71a1e87cc45.png)
Fast Blue RR Salt
![Fast Blue RR Salt Preweighed Capsules](/deepweb/assets/sigmaaldrich/product/structures/114/647/816c95a3-8172-4ea0-b156-e71a1e87cc45/640/816c95a3-8172-4ea0-b156-e71a1e87cc45.png)
Synonym(s):
4-Benzoylamino-2,5-dimethoxybenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 24
Linear Formula:
C15H14ClN3O3 · 1/2 ZnCl2
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing