Ethyl Orange sodium salt
Synonym(s):
4-(4-Diethylaminophenylazo)benzenesulfonic acid sodium salt
Linear Formula:
(C2H5)2NC6H4N=NC6H4SO3Na
Products
Product Category
Brand
Formula Weight
pH Value
Chemical Composition
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing