Skip to Content
Merck
Ethyl butyrate analytical standard

Ethyl butyrate

Ethyl butyrate analytical standard
Synonym(s):
Butyric acid ethyl ester
Linear Formula:
CH3CH2CH2C(O)OC2H5
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing