Skip to Content
Merck
Ethyl 4-oxo-1-piperidinecarboxylate 98%

Ethyl 4-oxo-1-piperidinecarboxylate

Ethyl 4-oxo-1-piperidinecarboxylate 98%
Synonym(s):
1-Carbethoxy-4-piperidone, 1-Ethoxycarbonyl-4-piperidone
Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing