Estradiol Impurity L
Synonym(s):
α-Estradiol, 1,3,5(10)-Estratriene-3,17α-diol, 3,17α-Dihydroxy-1,3,5(10)-estratriene, Epiestradiol
Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing