
Dyphylline

Synonym(s):
7-(2,3-Dihydroxypropyl)theophylline, 7-(2,3-Dihydroxypropyl)-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione, Diprophylline, Dyphylline
Empirical Formula (Hill Notation):
C10H14N4O4
CAS Number:
Molecular Weight:
254.24
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing