Skip to Content
Merck
Doxepin hydrochloride British Pharmacopoeia (BP) Reference Standard

Doxepin hydrochloride

Doxepin hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride
Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
EC Number:

Products

Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Chemical Composition
Application
API Family
Reference Material Type
Special Grade