![DL-Panthenol 99%](/deepweb/assets/sigmaaldrich/product/structures/165/473/f97e9f6e-7f13-4c8b-ba7a-415cd3dd21eb/640/f97e9f6e-7f13-4c8b-ba7a-415cd3dd21eb.png)
DL-Panthenol
![DL-Panthenol 99%](/deepweb/assets/sigmaaldrich/product/structures/165/473/f97e9f6e-7f13-4c8b-ba7a-415cd3dd21eb/640/f97e9f6e-7f13-4c8b-ba7a-415cd3dd21eb.png)
Synonym(s):
(±)-α,γ-Dihydroxy-N-(3-hydroxypropyl)-β,β-dimethylbutyramide, (±)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, DL-Pantothenyl alcohol
Linear Formula:
HOCH2C(CH3)2CH(OH)CONH(CH2)3OH
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing