![DL-Methionine sulfoxide ≥98.5% (NT)](/deepweb/assets/sigmaaldrich/product/structures/231/315/caf75dd9-ca09-4b0b-b715-b14d32299a08/640/caf75dd9-ca09-4b0b-b715-b14d32299a08.png)
DL-Methionine sulfoxide
![DL-Methionine sulfoxide ≥98.5% (NT)](/deepweb/assets/sigmaaldrich/product/structures/231/315/caf75dd9-ca09-4b0b-b715-b14d32299a08/640/caf75dd9-ca09-4b0b-b715-b14d32299a08.png)
Linear Formula:
CH3SOCH2CH2CH(NH2)COOH
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing