
DL-Lactaldehyde solution

Synonym(s):
2-Hydroxypropanal solution, 2-Hydroxypropionaldehyde solution
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Products
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing