Di-tert-butylchlorosilane
Synonym(s):
Chloro-di-tert-butylsilane
Linear Formula:
[(CH3)3C]2SiHCl
CAS Number:
Molecular Weight:
178.77
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing