
Disopyramide phosphate

Synonym(s):
Disopyramide phosphate salt, α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide
Linear Formula:
C21H29N3O · H3PO4
CAS Number:
Molecular Weight:
437.47
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2