Skip to Content
Merck
di-Sodium hydrogen phosphate heptahydrate EMPROVE® EXPERT, DAC, USP

di-Sodium hydrogen phosphate heptahydrate

di-Sodium hydrogen phosphate heptahydrate EMPROVE® EXPERT, DAC, USP
Synonym(s):
Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate
Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group