![(+)-Ipc2B(allyl)borane solution 1 M in pentane](/deepweb/assets/sigmaaldrich/product/structures/355/760/f2888777-4b75-427f-b588-bd2b105751d2/640/f2888777-4b75-427f-b588-bd2b105751d2.png)
(+)-Ipc2B(allyl)borane solution
![(+)-Ipc2B(allyl)borane solution 1 M in pentane](/deepweb/assets/sigmaaldrich/product/structures/355/760/f2888777-4b75-427f-b588-bd2b105751d2/640/f2888777-4b75-427f-b588-bd2b105751d2.png)
Synonym(s):
(+)-B-Allyldiisopinocampheylborane solution
Linear Formula:
(C10H17)2BCH2CH=CH2
CAS Number:
Molecular Weight:
326.37
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing