![Dihydroxyacetone Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/422/422/094b8df7-4d47-42e4-84f9-db41c4555a3d/640/094b8df7-4d47-42e4-84f9-db41c4555a3d.jpg)
Dihydroxyacetone
![Dihydroxyacetone Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/422/422/094b8df7-4d47-42e4-84f9-db41c4555a3d/640/094b8df7-4d47-42e4-84f9-db41c4555a3d.jpg)
Synonym(s):
Dihydroxyacetone, 1,3-Dihydroxy-2-propanone, 1,3-Dihydroxyacetone, DHA, Glycerone, NSC 24343
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3